Assay Details
Target Gene Details
Entrez Gene ID: | 57062 |
Gene Name: | DEAD-box helicase 24 |
Gene Aliases: |
- |
Location: |
Chr.14:94050922-94081212 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDX24 | NM_020414.3 | NP_065147.1 | ||
AB209382.1 | BAD92619.1 | |||
AF134475.1 | AAP97252.1 | |||
AF145022.1 | AAP97274.1 | |||
AF214731.1 | AAG02169.1 | |||
AK025162.1 | BAB15079.1 | |||
AK294613.1 | ||||
AK296726.1 | ||||
AK297232.1 | ||||
AK304504.1 | ||||
AL136886.1 | CAB66820.1 | |||
BC008847.2 | AAH08847.1 | |||
BC009406.2 | AAH09406.1 | |||
BC096826.1 | AAH96826.1 | |||
BX537533.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34067 | Chr.14:94024355 - 94160851 on Build GRCh38 | Loss | IFI27L1 OTUB2 IFI27 IFI27L2 DDX24 |
esv3635373 | Chr.14:94040151 - 94173989 on Build GRCh38 | Gain | IFI27L1 OTUB2 IFI27 IFI27L2 DDX24 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |