Assay Details
Target Gene Details
Entrez Gene ID: | 4857 |
Gene Name: | NOVA alternative splicing regulator 1 |
Gene Aliases: |
Nova-1 |
Location: |
Chr.14:26445883-26597754 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NOVA1 | NM_002515.2 | NP_002506.2 | ||
NM_006489.2 | NP_006480.2 | |||
XM_011536800.2 | XP_011535102.1 | |||
XM_017021342.1 | XP_016876831.1 | |||
XM_017021343.1 | XP_016876832.1 | |||
XM_017021344.1 | XP_016876833.1 | |||
XM_017021345.1 | XP_016876834.1 | |||
XM_017021346.1 | XP_016876835.1 | |||
XM_017021347.1 | XP_016876836.1 | |||
XM_017021348.1 | XP_016876837.1 | |||
XM_017021349.1 | XP_016876838.1 | |||
AK226114.1 | ||||
AK291022.1 | ||||
AK291468.1 | ||||
AK301526.1 | ||||
BC075038.2 | AAH75038.1 | |||
BC075039.2 | AAH75039.1 | |||
U04840.1 | AAA16022.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv564107 | Chr.14:26429752 - 26489004 on Build GRCh38 | Loss | NOVA1 |
nsv1052199 | Chr.14:26334093 - 26735737 on Build GRCh38 | Loss | NOVA1-AS1 NOVA1 |
esv3633850 | Chr.14:26411340 - 26535541 on Build GRCh38 | Gain | NOVA1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |