Assay Details
Target Gene Details
Entrez Gene ID: | 10202 |
Gene Name: | dehydrogenase/reductase 2 |
Gene Aliases: |
HEP27, SDR25C1 |
Location: |
Chr.14:23630115-23645639 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DHRS2 | NM_001318835.1 | NP_001305764.1 | ||
XM_005267249.1 | XP_005267306.1 | |||
XM_006720001.3 | XP_006720064.1 | |||
XM_011536339.2 | XP_011534641.1 | |||
AK309975.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv274985 | Chr.14:23631250 - 23634912 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)