Assay Details
Target Gene Details
Entrez Gene ID: | 54331 |
Gene Name: | G protein subunit gamma 2 |
Gene Aliases: |
- |
Location: |
Chr.14:51847131-51969800 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNG2 | XM_006720173.2 | XP_006720236.1 | ||
XM_017021377.1 | XP_016876866.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952840 | Chr.14:51846583 - 51949982 on Build GRCh38 | Duplication | GNG2 LOC102723604 |
nsv564842 | Chr.14:51803013 - 51867871 on Build GRCh38 | Loss | LOC101927598 GNG2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |