Assay Details
Target Gene Details
Entrez Gene ID: | 51528 |
Gene Name: | JNK1/MAPK8-associated membrane protein |
Gene Aliases: |
C14orf100, CDA06, HSPC213, HSPC327, JAMP |
Location: |
Chr.14:59484443-59505407 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JKAMP | NM_001098625.2 | NP_001092095.1 | ||
NM_001284201.1 | NP_001271130.1 | |||
NM_001284202.1 | NP_001271131.1 | |||
NM_001284203.1 | NP_001271132.1 | |||
NM_001284204.1 | NP_001271133.1 | |||
NM_016475.4 | NP_057559.2 | |||
XM_006720171.3 | XP_006720234.1 | |||
XM_017021366.1 | XP_016876855.1 | |||
AF151047.1 | AAF36133.1 | |||
AF161445.1 | AAF29005.1 | |||
AF212245.1 | AAG41781.1 | |||
AF529304.1 | AAP85636.1 | |||
AK021683.1 | BAB13874.1 | |||
AK027591.1 | BAB55216.1 | |||
AK056145.1 | ||||
AK128628.1 | 1 | 1199 | ||
AK298209.1 | ||||
AL136696.1 | CAB66631.1 | |||
BC005198.1 | AAH05198.1 | |||
BC010359.1 | AAH10359.1 | |||
BU928810.1 | ||||
BX161501.1 | CAD61945.1 | |||
CR457163.1 | CAG33444.1 | |||
CR533510.1 | CAG38541.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |