Assay Details
Target Gene Details
Entrez Gene ID: | 2957 |
Gene Name: | general transcription factor IIA subunit 1 |
Gene Aliases: |
TF2A1, TFIIA, TFIIA-42, TFIIAL |
Location: |
Chr.14:81175452-81221231 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GTF2A1 | NM_001278940.1 | NP_001265869.1 | ||
NM_015859.3 | NP_056943.1 | |||
NM_201595.2 | NP_963889.1 | |||
BC107155.2 | AAI07156.1 | |||
BC107156.1 | AAI07157.1 | |||
D14886.1 | BAA03603.1 | |||
D14887.1 | BAA03604.1 | |||
X75383.1 | CAA53151.1 | |||
X77225.1 | CAA54442.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3581341 | Chr.14:81178660 - 81196157 on Build GRCh38 | Loss | GTF2A1 |
nsv832841 | Chr.14:81121978 - 81277577 on Build GRCh38 | Gain | TSHR LOC101928504 SNORA79 GTF2A1 STON2 |
esv3635076 | Chr.14:80715257 - 81672373 on Build GRCh38 | Gain | LINC01467 TSHR LOC101928504 SNORA79 CEP128 GTF2A1 LOC100506700 SEL1L STON2 |
esv3581342 | Chr.14:81187422 - 81196157 on Build GRCh38 | Loss | GTF2A1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |