Assay Details
Target Gene Details
Entrez Gene ID: | 122809 |
Gene Name: | suppressor of cytokine signaling 4 |
Gene Aliases: |
SOCS7 |
Location: |
Chr.14:55027126-55049489 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SOCS4 | NM_080867.2 | NP_543143.1 | ||
NM_199421.1 | NP_955453.1 | |||
XM_011536425.1 | XP_011534727.1 | |||
XM_011536426.1 | XP_011534728.1 | |||
AK124083.1 | ||||
BC060790.1 | AAH60790.1 | |||
BX161418.1 | CAD61893.1 | |||
BX648734.1 | ||||
CD109883.1 | ||||
CR933659.1 | CAI45958.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |