Assay Details
Target Gene Details
Entrez Gene ID: | 10082 |
Gene Name: | glypican 6 |
Gene Aliases: |
OMIMD1 |
Location: |
Chr.13:93226825-94408020 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPC6 | NM_005708.3 | NP_005699.1 | ||
AF105267.1 | AAD55749.1 | |||
AF111178.1 | AAD31392.1 | |||
AK074741.1 | ||||
AK130013.1 | ||||
AK290144.1 | ||||
AK304338.1 | ||||
AY358462.1 | AAQ88827.1 | |||
BC106947.2 | AAI06948.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3582694 | Chr.13:91877905 - 93587318 on Build GRCh38 | Gain | GPC5 LINC00363 GPC6 GPC5-AS1 MIR548AS GPC5-AS2 |
esv3892424 | Chr.13:92937463 - 94196357 on Build GRCh38 | Gain | GPC6-AS1 LINC00363 GPC6-AS2 GPC6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |