Assay Details
Target Gene Details
Entrez Gene ID: | 337867 |
Gene Name: | UBA domain containing 2 |
Gene Aliases: |
PHGDHL1 |
Location: |
Chr.13:99200425-99386499 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBAC2 | NM_001144072.1 | NP_001137544.1 | ||
NM_177967.3 | NP_808882.1 | |||
NR_026644.1 | ||||
XM_006719948.3 | XP_006720011.1 | |||
XM_011521082.2 | XP_011519384.1 | |||
XM_011521083.2 | XP_011519385.1 | |||
AK054563.1 | BAB70757.1 | |||
AK075419.1 | BAC11609.1 | |||
AK075516.1 | BAC11665.1 | |||
AK124404.1 | ||||
AK129861.1 | ||||
AK290342.1 | ||||
AK300908.1 | ||||
BC121138.1 | ||||
BC121139.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892433 | Chr.13:98843569 - 99243711 on Build GRCh38 | Gain | UBAC2 DOCK9-AS2 DOCK9 UBAC2-AS1 |
nsv1050094 | Chr.13:98974062 - 99323969 on Build GRCh38 | Gain | UBAC2 DOCK9-AS2 DOCK9 GPR18 UBAC2-AS1 GPR183 |
esv3582749 | Chr.13:99137911 - 99253291 on Build GRCh38 | Gain | UBAC2 UBAC2-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |