Assay Details
Target Gene Details
Entrez Gene ID: | 253512 |
Gene Name: | solute carrier family 25 member 30 |
Gene Aliases: |
KMCP1 |
Location: |
Chr.13:45393316-45434016 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A30 | XM_017020523.1 | XP_016876012.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv223e201 | Chr.13:45377870 - 46464067 on Build GRCh38 | Deletion | TPT1-AS1 SLC25A30-AS1 ZC3H13 SPERT CPB2-AS1 KIAA0226L SLC25A30 COG3 ERICH6B LINC00563 LRRC63 CPB2 SIAH3 LCP1 LINC01198 LINC01055 |
nsv510579 | Chr.13:45305226 - 45424938 on Build GRCh38 | Deletion | TPT1-AS1 SLC25A30 TPT1 SLC25A30-AS1 SNORA31 |
nsv455875 | Chr.13:45415321 - 45479409 on Build GRCh38 | Loss | SLC25A30 COG3 SLC25A30-AS1 |
esv3631957 | Chr.13:45413790 - 45431769 on Build GRCh38 | Loss | SLC25A30 SLC25A30-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |