Assay Details
Target Gene Details
Entrez Gene ID: | 267012 |
Gene Name: | D-amino acid oxidase activator |
Gene Aliases: |
LG72, SG72 |
Location: |
Chr.13:105465867-105491034 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DAOA | NM_001161812.1 | NP_001155284.1 | ||
NM_001161814.1 | NP_001155286.1 | |||
NM_172370.4 | NP_758958.3 | |||
XM_005254042.1 | XP_005254099.1 | |||
AY138546.1 | AAN08432.1 | |||
AY138547.1 | AAN08433.1 | |||
AY170469.2 | AAO12727.1 | |||
AY170470.2 | AAO12728.1 | |||
AY170471.1 | AAO12729.1 | |||
AY223901.1 | AAO73604.1 | |||
BC121091.1 | ||||
DQ343761.1 | ||||
DQ357223.1 | ||||
DQ386869.1 | ||||
DQ386870.1 |
Target Gene Details
Entrez Gene ID: | 282706 |
Gene Name: | DAOA antisense RNA 1 |
Gene Aliases: |
DAOA-AS, DAOAAS, G30 |
Location: |
Chr.13:105459055-105505681 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DAOA-AS1 | NR_040247.1 | |||
AY138548.1 | AAN08434.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv275034 | Chr.13:105478789 - 105480809 on Build GRCh38 | Gain+Loss |
![]() ![]() |
nsv562996 | Chr.13:104426430 - 105664629 on Build GRCh38 | Gain |
![]() ![]() |
nsv1048036 | Chr.13:105463559 - 105534128 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)