Assay Details
Target Gene Details
Entrez Gene ID: | 81575 |
Gene Name: | apolipoprotein L domain containing 1 |
Gene Aliases: |
VERGE |
Location: |
Chr.12:12725917-12791466 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOLD1 | NM_001130415.1 | NP_001123887.1 | ||
BC042478.1 | AAH42478.1 | |||
DC413327.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2745567 | Chr.12:12246086 - 13092998 on Build GRCh38 | Deletion | MIR613 GPRC5A LOC100506314 HTR7P1 FAM234B DDX47 GPR19 RPL13AP20 BORCS5 LOH12CR2 MANSC1 HEBP1 LRP6 APOLD1 CDKN1B LOC105369662 MIR614 GSG1 DUSP16 CREBL2 GPRC5D |
nsv1046355 | Chr.12:11501836 - 12890185 on Build GRCh38 | Loss | MIR613 MIR1244-4 DDX47 GPR19 RPL13AP20 BORCS5 LOH12CR2 LINC01252 MANSC1 LRP6 BCL2L14 APOLD1 CDKN1B LOC105369662 DUSP16 ETV6 LOC100506248 CREBL2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |