Assay Details
Target Gene Details
Entrez Gene ID: | 8739 |
Gene Name: | harakiri, BCL2 interacting protein |
Gene Aliases: |
DP5, HARAKIRI |
Location: |
Chr.12:116856144-116881427 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HRK | NM_003806.2 | NP_003797.1 | ||
NR_073189.1 | ||||
XM_017020146.1 | XP_016875635.1 | |||
BE674890.1 | ||||
D83699.1 | ||||
U76376.1 | AAC34931.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv898 | Chr.12:116849419 - 116884372 on Build GRCh38 | Insertion | RNFT2 HRK |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |