Assay Details
Target Gene Details
Entrez Gene ID: | 144608 |
Gene Name: | chromosome 12 open reading frame 60 |
Gene Aliases: |
- |
Location: |
Chr.12:14567991-14823858 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C12orf60 | XM_017018871.1 | XP_016874360.1 | ||
XM_017018872.1 | XP_016874361.1 |
Target Gene Details
Entrez Gene ID: | 105369669 |
Gene Name: | uncharacterized LOC105369669 |
Gene Aliases: |
- |
Location: |
Chr.12:14746666-14770662 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105369669 | XM_011520896.2 | XP_011519198.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)