Assay Details
Target Gene Details
Entrez Gene ID: | 6128 |
Gene Name: | ribosomal protein L6 |
Gene Aliases: |
L6, SHUJUN-2, TAXREB107, TXREB1 |
Location: |
Chr.12:112405181-112418850 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RPL6 | NM_001320141.1 | NP_001307070.1 | ||
XM_017019781.1 | XP_016875270.1 | |||
XM_017019782.1 | XP_016875271.1 | |||
BG722198.1 | ||||
HY038019.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3580466 | Chr.12:112403584 - 112419956 on Build GRCh38 | Loss | RPL6 PTPN11 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |