Assay Details
Target Gene Details
Entrez Gene ID: | 283314 |
Gene Name: | C1RL antisense RNA 1 |
Gene Aliases: |
MATL2963 |
Location: |
Chr.12:7108308-7121851 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C1RL-AS1 | NR_026947.1 | 8 | 4101 | |
AK091792.1 | ||||
AL833364.1 | 8 | 4101 |