Assay Details
Target Gene Details
Entrez Gene ID: | 8655 |
Gene Name: | dynein light chain LC8-type 1 |
Gene Aliases: |
DLC1, DLC8, DNCL1, DNCLC1, LC8, LC8a, PIN, hdlc1 |
Location: |
Chr.12:120469857-120498495 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DYNLL1 | NM_001037494.1 | NP_001032583.1 | ||
BG474448.1 | ||||
CB992195.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509482 | Chr.12:120435117 - 120495113 on Build GRCh38 | Insertion | GATC SRSF9 DYNLL1 TRIAP1 COX6A1 NRAV |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |