Assay Details
Target Gene Details
Entrez Gene ID: | 1108 |
Gene Name: | chromodomain helicase DNA binding protein 4 |
Gene Aliases: |
CHD-4, Mi-2b, Mi2-BETA |
Location: |
Chr.12:6570082-6607439 on Build GRCh38 |
Assay Gene Location: | Within Intron 36 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509452 | Chr.12:6566935 - 6653627 on Build GRCh38 | Insertion | ING4 CHD4 SCARNA11 ACRBP NOP2 LPAR5 |
nsv826210 | Chr.12:6561863 - 6612310 on Build GRCh38 | Gain | CHD4 SCARNA11 NOP2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |