Assay Details
Target Gene Details
Entrez Gene ID: | 2059 |
Gene Name: | epidermal growth factor receptor pathway substrate 8 |
Gene Aliases: |
DFNB102 |
Location: |
Chr.12:15620141-15789576 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPS8 | NM_004447.5 | NP_004438.3 | ||
XM_011520605.2 | XP_011518907.1 | |||
AK291777.1 | ||||
AK292931.1 | ||||
AK301834.1 | ||||
AK304860.1 | ||||
AK316134.1 | ||||
AK316239.1 | ||||
BC030010.2 | AAH30010.1 | |||
U12535.1 | AAA62280.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |