Assay Details
Target Gene Details
Entrez Gene ID: | 100287944 |
Gene Name: | uncharacterized LOC100287944 |
Gene Aliases: |
- |
Location: |
Chr.12:106496410-106774831 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100287944 | NR_040246.1 | |||
DA250223.1 |
Target Gene Details
Entrez Gene ID: | 5992 |
Gene Name: | regulatory factor X4 |
Gene Aliases: |
NYD-SP10 |
Location: |
Chr.12:106582907-106762804 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RFX4 | NM_213594.2 | NP_998759.1 | ||
AB095366.1 | BAE48232.1 | |||
AK291445.1 | ||||
AY102009.1 | AAM52484.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832511 | Chr.12:106551876 - 106714050 on Build GRCh38 | Gain | LOC100505978 LOC100287944 RFX4 |
nsv560090 | Chr.12:106541358 - 106830191 on Build GRCh38 | Gain | LOC100505978 LOC100287944 RIC8B RFX4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |