Assay Details
Target Gene Details
Entrez Gene ID: | 60488 |
Gene Name: | mitochondrial ribosomal protein S35 |
Gene Aliases: |
HDCMD11P, MDS023, MRP-S28, MRPS28 |
Location: |
Chr.12:27710773-27756304 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MRPS35 | NM_001190864.1 | NP_001177793.1 | ||
NM_021821.3 | NP_068593.2 | |||
AF068296.1 | AAF65185.1 | |||
AF182422.1 | AAG14958.1 | |||
AK075378.1 | BAC11579.1 | |||
AK075515.1 | BAC11664.1 | |||
AL512733.1 | CAC21665.1 | |||
BC015862.1 | ||||
BC017086.1 | AAH17086.1 | |||
BC028346.2 | AAH28346.2 | |||
BC063515.1 | AAH63515.1 | |||
BC109372.1 | AAI09373.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2745664 | Chr.12:27017516 - 27820117 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv469178 | Chr.12:27695850 - 27762012 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)