Assay Details
Target Gene Details
Entrez Gene ID: | 57379 |
Gene Name: | activation-induced cytidine deaminase |
Gene Aliases: |
AID, ARP2, CDA2, HEL-S-284, HIGM2 |
Location: |
Chr.12:8602166-8612970 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AICDA | NM_020661.2 | NP_065712.1 | ||
XM_011520772.2 | XP_011519074.1 | |||
XM_011520773.2 | XP_011519075.1 | |||
AB040431.1 | BAB12721.1 | |||
BC006296.2 | AAH06296.1 | |||
BX402063.2 | ||||
EU794587.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain | LOC105369649 KLRG1 M6PR LOC105376678 LOC105369728 MIR1244-3 CLEC2A OVOS KLRB1 KLRF2 MFAP5 LINC00612 AICDA CLEC4E A2M CLEC6A CLEC2B PZP CLEC12A LOC374443 PHC1 CLEC4D LOC642846 KLRF1 CLEC2D CD69 LOC101928030 LOC400002 LOC101930452 CLECL1 DDX12P A2MP1 A2M-AS1 RIMKLB LOC100506159 LINC00987 LOC728715 A2ML1 |
nsv557342 | Chr.12:8446801 - 8621931 on Build GRCh38 | Loss | AICDA CLEC4D CLEC4E CLEC6A |
dgv1362n100 | Chr.12:8296504 - 8639973 on Build GRCh38 | Gain | AICDA CLEC4D CLEC4E CLEC6A LINC00937 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |