Assay Details
Target Gene Details
Entrez Gene ID: | 5939 |
Gene Name: | RNA binding motif single stranded interacting protein 2 |
Gene Aliases: |
SCR3 |
Location: |
Chr.12:56521812-56596196 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RBMS2 | NM_002898.3 | NP_002889.1 | ||
XM_005269059.4 | XP_005269116.1 | |||
XM_005269060.4 | XP_005269117.1 | |||
XM_005269061.3 | XP_005269118.1 | |||
XM_005269066.4 | XP_005269123.1 | |||
XM_006719541.3 | XP_006719604.1 | |||
XM_006719542.3 | XP_006719605.1 | |||
XM_006719543.3 | XP_006719606.1 | |||
XM_006719544.3 | XP_006719607.1 | |||
XM_011538638.2 | XP_011536940.1 | |||
XM_011538639.2 | XP_011536941.1 | |||
XM_011538640.2 | XP_011536942.1 | |||
AK293315.1 | ||||
AK299580.1 | ||||
AK304540.1 | ||||
AK308885.1 | ||||
BC027863.1 | AAH27863.1 | |||
BC072679.1 | AAH72679.1 | |||
D28483.1 | BAA05842.1 | |||
DC382164.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss | SNORD59A MIR6758 TIMELESS DDIT3 GPR182 ARHGAP9 GLI1 TAC3 PAN2 HSD17B6 SNORA105C MBD6 B4GALNT1 ANKRD52 HBCBP RBMS2 CS MARS MIR1228 SLC39A5 NAB2 DTX3 LRP1-AS MIR616 STAT6 INHBC BAZ2A NDUFA4L2 SLC26A10 SDR9C7 CNPY2 NACA SNORD59B NABP2 NXPH4 SHMT2 COQ10A KIF5A PIP4K2C NEMP1 LRP1 APOF INHBE OS9 IL23A ZBTB39 DCTN2 ATP5B RDH16 STAT2 STAC3 R3HDM2 RNF41 GLS2 LOC101927583 SPRYD4 PTGES3 ARHGEF25 PRIM1 MYO1A MIP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |