Assay Details
Target Gene Details
Entrez Gene ID: | 8630 |
Gene Name: | hydroxysteroid 17-beta dehydrogenase 6 |
Gene Aliases: |
HSE, RODH, SDR9C6 |
Location: |
Chr.12:56752449-56787790 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HSD17B6 | NM_003725.3 | NP_003716.2 | ||
XM_005269207.1 | XP_005269264.1 | |||
XM_005269208.1 | XP_005269265.1 | |||
XM_005269209.1 | XP_005269266.1 | |||
XM_006719672.1 | XP_006719735.1 | |||
XM_011538925.1 | XP_011537227.1 | |||
XM_011538926.1 | XP_011537228.1 | |||
XM_011538927.1 | XP_011537229.1 | |||
AF223225.1 | AAF81017.1 | |||
AK313423.1 | ||||
BC020710.1 | AAH20710.1 | |||
DA086202.1 | ||||
U89281.1 | AAB88252.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3629600 | Chr.12:56762971 - 56780702 on Build GRCh38 | Loss |
![]() |
esv2667772 | Chr.12:56763048 - 56780839 on Build GRCh38 | Deletion |
![]() |
nsv826382 | Chr.12:56757680 - 56814823 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)