Assay Details
Target Gene Details
Entrez Gene ID: | 92797 |
Gene Name: | DNA helicase B |
Gene Aliases: |
DHB, hDHB |
Location: |
Chr.12:66302545-66343643 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HELB | NM_033647.4 | NP_387467.2 | ||
NR_135080.1 | ||||
NR_135081.1 | ||||
XM_005269234.2 | XP_005269291.1 | |||
XM_017020230.1 | XP_016875719.1 | |||
AF319995.1 | AAM73554.1 | |||
AK290894.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892212 | Chr.12:66302630 - 66471921 on Build GRCh38 | Loss | GRIP1 HELB |
esv3629801 | Chr.12:66277937 - 66308122 on Build GRCh38 | Gain | HELB |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |