Assay Details
Target Gene Details
Entrez Gene ID: | 2026 |
Gene Name: | enolase 2 |
Gene Aliases: |
HEL-S-279, NSE |
Location: |
Chr.12:6914450-6923696 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENO2 | NM_001975.2 | NP_001966.1 | ||
AK124656.1 | ||||
AK290525.1 | ||||
AK295220.1 | ||||
AK312402.1 | ||||
BC002745.2 | AAH02745.1 | |||
BT007383.1 | AAP36047.1 | |||
CR536582.1 | CAG38819.1 | |||
EU794607.1 | ||||
M22349.1 | AAB59554.1 | |||
M36768.1 | AAA52388.1 | |||
X13120.1 | CAA31512.1 | |||
X14327.1 | CAA32505.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1047373 | Chr.12:6902891 - 7055754 on Build GRCh38 | Gain | C12orf57 LRRC23 EMG1 PHB2 LOC105369635 SCARNA12 RNU7-1 MIR141 PTPN6 ENO2 MIR200C LPCAT3 ATN1 |
nsv509453 | Chr.12:6896530 - 7007852 on Build GRCh38 | Insertion | C12orf57 LRRC23 EMG1 PHB2 LOC105369635 SCARNA12 RNU7-1 MIR141 PTPN6 ENO2 MIR200C LPCAT3 ATN1 |
nsv557261 | Chr.12:6839304 - 6924660 on Build GRCh38 | Loss | LRRC23 USP5 TPI1 GNB3 P3H3 RPL13P5 LOC105369632 DSTNP2 CDCA3 ENO2 SPSB2 ATN1 |
nsv832324 | Chr.12:6780297 - 6932306 on Build GRCh38 | Loss | GPR162 LRRC23 USP5 CD4 GNB3 RPL13P5 LOC105369632 CDCA3 SPSB2 TPI1 P3H3 DSTNP2 ENO2 ATN1 |
nsv1035811 | Chr.12:6876316 - 6994911 on Build GRCh38 | Gain | C12orf57 LRRC23 EMG1 PHB2 LOC105369635 RPL13P5 LOC105369632 SCARNA12 RNU7-1 MIR141 PTPN6 DSTNP2 ENO2 MIR200C LPCAT3 ATN1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |