Assay Details
Target Gene Details
Entrez Gene ID: | 80724 |
Gene Name: | acyl-CoA dehydrogenase family member 10 |
Gene Aliases: |
- |
Location: |
Chr.12:111686053-111757107 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACAD10 | NM_001136538.1 | NP_001130010.1 | ||
NM_025247.5 | NP_079523.3 | |||
AK092356.1 | BAC03869.1 | |||
AK310923.1 | ||||
AL832043.1 | ||||
AY323912.1 | AAQ88260.1 | |||
BC015056.1 | AAH15056.1 | |||
BC126358.1 | ||||
DA146693.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3630763 | Chr.12:111684168 - 111696825 on Build GRCh38 | Gain |
![]() ![]() |
esv3630762 | Chr.12:111679417 - 111731317 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)