Assay Details
Target Gene Details
Entrez Gene ID: | 100286844 |
Gene Name: | BCDIN3D antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:49828543-49841154 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCDIN3D-AS1 | NR_027499.1 | |||
NR_027500.1 | ||||
NR_027501.1 | ||||
AI193197.1 | ||||
AK055142.1 | ||||
CB153643.1 | ||||
DA122615.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522972 | Chr.12:49802611 - 50017048 on Build GRCh38 | Gain | AQP5 LOC101927292 BCDIN3D LOC101927318 AQP6 BCDIN3D-AS1 AQP2 NCKAP5L LOC283332 RACGAP1 FAIM2 |