Assay Details
Target Gene Details
Entrez Gene ID: | 51228 |
Gene Name: | glycolipid transfer protein |
Gene Aliases: |
- |
Location: |
Chr.12:109850943-109880488 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLTP | NM_016433.3 | NP_057517.1 | ||
XM_011538434.2 | XP_011536736.1 | |||
AF209704.1 | AAF33210.1 | |||
AK313457.1 | ||||
AY372530.1 | AAR85984.1 | |||
AY372531.1 | AAR85985.1 | |||
AY372532.1 | AAR87373.1 | |||
BC009932.2 | AAH09932.1 | |||
JN039379.1 | ||||
JN052207.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |