Assay Details
Target Gene Details
Entrez Gene ID: | 56901 |
Gene Name: | NDUFA4, mitochondrial complex associated like 2 |
Gene Aliases: |
NUOMS |
Location: |
Chr.12:57234903-57240762 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFA4L2 | XM_005269033.2 | XP_005269090.1 | ||
AK094275.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss | CNPY2 SNORA105C SPRYD4 MYO1A STAT6 GPR182 BAZ2A B4GALNT1 GLS2 OS9 RBMS2 NEMP1 SHMT2 GLI1 SNORD59B MIR6758 NABP2 ANKRD52 STAC3 INHBE NXPH4 MBD6 COQ10A LOC101927583 TIMELESS APOF NACA CS IL23A MIR1228 R3HDM2 PAN2 TAC3 RNF41 KIF5A SNORD59A LRP1-AS SLC39A5 PIP4K2C HSD17B6 DDIT3 MIP SDR9C7 NDUFA4L2 SLC26A10 INHBC HBCBP PRIM1 PTGES3 ARHGAP9 ZBTB39 STAT2 NAB2 ARHGEF25 ATP5B DTX3 LRP1 RDH16 MARS MIR616 DCTN2 |
nsv519293 | Chr.12:57189889 - 57243810 on Build GRCh38 | Loss | SHMT2 STAC3 LRP1 NXPH4 MIR1228 NDUFA4L2 |
nsv952189 | Chr.12:57127118 - 57245617 on Build GRCh38 | Deletion | SHMT2 STAC3 LRP1 NXPH4 MIR1228 LRP1-AS NDUFA4L2 |
dgv96n21 | Chr.12:57139199 - 57243810 on Build GRCh38 | Loss | SHMT2 STAC3 LRP1 NXPH4 MIR1228 LRP1-AS NDUFA4L2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |