Assay Details
Target Gene Details
Entrez Gene ID: | 79023 |
Gene Name: | nucleoporin 37 |
Gene Aliases: |
p37 |
Location: |
Chr.12:102074189-102120139 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NUP37 | NM_024057.3 | NP_076962.2 | ||
AF514994.1 | AAM76705.1 | |||
AK026271.1 | BAB15422.1 | |||
AK296733.1 | ||||
AK310619.1 | ||||
BC000861.2 | AAH00861.2 | |||
BM995521.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)