Assay Details
Target Gene Details
Entrez Gene ID: | 4257 |
Gene Name: | microsomal glutathione S-transferase 1 |
Gene Aliases: |
GST12, MGST, MGST-I |
Location: |
Chr.12:16347142-16377189 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MGST1 | NM_001260511.1 | NP_001247440.1 | ||
NM_001260512.1 | NP_001247441.1 | |||
NM_001267598.1 | NP_001254527.1 | |||
NM_020300.4 | NP_064696.1 | |||
NM_145791.2 | NP_665734.1 | |||
NM_145792.2 | NP_665735.1 | |||
NR_048545.1 | ||||
NR_048547.1 | ||||
XM_006719079.1 | XP_006719142.1 | |||
XM_011520673.1 | XP_011518975.1 | |||
XM_011520674.2 | XP_011518976.1 | |||
AA314967.1 | ||||
AK291148.1 | ||||
AK309707.1 | ||||
AV655242.1 | ||||
BC005923.1 | AAH05923.1 | |||
BE568125.1 | ||||
BG569388.1 | ||||
BG617746.1 | ||||
BG778603.1 | ||||
H49331.1 | ||||
R86750.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1049805 | Chr.12:16284461 - 16498558 on Build GRCh38 | Loss | MGST1 |
nsv1036638 | Chr.12:15912446 - 16358900 on Build GRCh38 | Loss | SLC15A5 MGST1 DERA |
esv3892149 | Chr.12:16266471 - 16354205 on Build GRCh38 | Loss | SLC15A5 MGST1 |
esv3892150 | Chr.12:16339581 - 16384735 on Build GRCh38 | Loss | MGST1 |
nsv832338 | Chr.12:16319300 - 16487163 on Build GRCh38 | Gain | MGST1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |