Assay Details
Target Gene Details
Entrez Gene ID: | 83591 |
Gene Name: | THAP domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.12:71663897-71680648 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
THAP2 | NM_031435.3 | NP_113623.1 | ||
AK313452.1 | ||||
AL136607.1 | CAB66542.1 | |||
BC008358.1 | AAH08358.1 | |||
CR533485.1 | CAG38516.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832451 | Chr.12:71584552 - 71806081 on Build GRCh38 | Loss | TMEM19 RAB21 THAP2 LGR5 ZFC3H1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |