Assay Details
Target Gene Details
Entrez Gene ID: | 775 |
Gene Name: | calcium voltage-gated channel subunit alpha1 C |
Gene Aliases: |
CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS |
Location: |
Chr.12:1969677-2697949 on Build GRCh38 |
Assay Gene Location: | Within Intron 45 |
Target Gene Details
Entrez Gene ID: | 100874235 |
Gene Name: | CACNA1C antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.12:2668500-2672220 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CACNA1C-AS2 | NR_046579.1 | |||
AI219155.1 | ||||
AI797712.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1047874 | Chr.12:2577599 - 2807322 on Build GRCh38 | Gain | CACNA1C-AS1 CACNA1C-AS2 FKBP4 CACNA1C LOC283440 |
nsv832308 | Chr.12:2576562 - 2690062 on Build GRCh38 | Loss | CACNA1C-AS1 CACNA1C-AS2 CACNA1C |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |