Assay Details
Target Gene Details
Entrez Gene ID: | 84260 |
Gene Name: | trichoplein keratin filament binding |
Gene Aliases: |
TpMs |
Location: |
Chr.12:109880667-109918069 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TCHP | XM_011538837.2 | XP_011537139.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)