Assay Details
Target Gene Details
Entrez Gene ID: | 22927 |
Gene Name: | hyaluronan binding protein 4 |
Gene Aliases: |
IHABP-4, IHABP4, Ki-1/57, SERBP1L |
Location: |
Chr.9:96450155-96491336 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HABP4 | NM_014282.3 | NP_055097.2 | ||
XM_005251812.3 | XP_005251869.1 | |||
XM_017014484.1 | XP_016869973.1 | |||
AF241831.1 | AAF62546.1 | |||
BC018788.1 | AAH18788.1 | |||
BQ962055.1 | ||||
DR006985.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv824998 | Chr.9:96394002 - 96467870 on Build GRCh38 | Gain | ZNF367 HABP4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |