Assay Details
Target Gene Details
Entrez Gene ID: | 50808 |
Gene Name: | adenylate kinase 3 |
Gene Aliases: |
AK3L1, AK6, AKL3L, AKL3L1, FIX |
Location: |
Chr.9:4709556-4742043 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AK3 | NM_001199852.1 | NP_001186781.1 | ||
NM_001199853.1 | NP_001186782.1 | |||
NM_001199855.1 | NP_001186784.1 | |||
NM_001199856.1 | NP_001186785.1 | |||
NM_016282.3 | NP_057366.2 | |||
AB021870.1 | BAA87913.1 | |||
AF086913.1 | AAP97143.1 | |||
AF100785.1 | AAP97224.1 | |||
AF183419.1 | AAG09688.1 | |||
AK001553.1 | BAA91753.1 | |||
AK001951.1 | BAA91996.1 | |||
AK027534.1 | BAB55183.1 | |||
AK098205.1 | ||||
AK124824.1 | ||||
AK298200.1 | ||||
BC013771.1 | AAH13771.1 | |||
BI550730.1 | ||||
BI601245.1 | ||||
DA570364.1 | ||||
DA718987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1015376 | Chr.9:4686223 - 4737890 on Build GRCh38 | Gain | CDC37L1 AK3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |