Assay Details
Target Gene Details
Entrez Gene ID: | 79695 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 12 |
Gene Aliases: |
CRCS1, GalNAc-T12 |
Location: |
Chr.9:98807699-98850081 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GALNT12 | NM_024642.4 | NP_078918.3 | ||
XM_006717287.1 | XP_006717350.1 | |||
XM_011519018.2 | XP_011517320.1 | |||
XM_011519020.1 | XP_011517322.1 | |||
XM_017015133.1 | XP_016870622.1 | |||
AB040673.1 | BAD93346.1 | |||
AB078146.1 | BAC07181.1 | |||
AJ132365.2 | CAC80100.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv614936 | Chr.9:98420416 - 98918098 on Build GRCh38 | Gain | GALNT12 ANKS6 GABBR2 |
esv3621214 | Chr.9:98651384 - 99637088 on Build GRCh38 | Gain | GALNT12 COL15A1 ANKS6 LOC101928438 ALG2 NAMA SEC61B TGFBR1 GABBR2 |
nsv825009 | Chr.9:98814132 - 98818055 on Build GRCh38 | Loss | GALNT12 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |