Assay Details
Target Gene Details
Entrez Gene ID: | 101929467 |
Gene Name: | LURAP1L antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:12699999-12814389 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LURAP1L-AS1 | NR_125775.1 | |||
BQ416907.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3891612 | Chr.9:12213879 - 13226946 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv1021844 | Chr.9:12356017 - 13444976 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv1016570 | Chr.9:12576095 - 12722388 on Build GRCh38 | Loss |
![]() ![]() |
esv3619718 | Chr.9:12621841 - 12766938 on Build GRCh38 | Gain |
![]() ![]() |
esv2422212 | Chr.9:12448789 - 13127739 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() |
esv2759671 | Chr.9:12235048 - 12784093 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv613613 | Chr.9:12641987 - 12773264 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)