Assay Details
Target Gene Details
Entrez Gene ID: | 8328 |
Gene Name: | growth factor independent 1B transcriptional repressor |
Gene Aliases: |
BDPLT17, ZNF163B |
Location: |
Chr.9:132945545-132991697 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GFI1B | NM_004188.6 | NP_004179.3 | ||
BC043371.1 | AAH43371.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509329 | Chr.9:132941822 - 132956074 on Build GRCh38 | Insertion | MIR548AW GFI1B TSC1 |
nsv831743 | Chr.9:132820292 - 132982548 on Build GRCh38 | Loss | MIR548AW GFI1B TSC1 AK8 SPACA9 |
esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion | MIR3689F RPL7A OLFM1 CACFD1 LOC101928193 LOC101928525 COL5A1 AK8 SPACA9 MIR3689B MIR4669 MIR3689D1 SURF2 TMEM8C REXO4 GTF3C5 SNORD24 MIR6877 GBGT1 FCN1 RALGDS DBH BRD3 CEL LOC101448202 SURF6 LOC105376306 WDR5 PPP1R26 MIR548AW DBH-AS1 SNORD36A SNORD36B MIR3689C MED22 MIR3689A ABO SNORD36C SURF1 ADAMTS13 MIR3689E RXRA C9orf62 TSC1 RNU6ATAC LINC00094 LOC100130548 GFI1B STKLD1 SNORD141A COL5A1-AS1 CELP FAM163B ADAMTSL2 LOC100996574 SLC2A6 SURF4 LOC401557 LCN1 PPP1R26-AS1 FCN2 OBP2B SARDH MIR3689D2 VAV2 C9orf116 MRPS2 |
nsv6745 | Chr.9:132929207 - 132958453 on Build GRCh38 | Insertion | MIR548AW GFI1B TSC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |