Assay Details
Target Gene Details
Entrez Gene ID: | 8328 |
Gene Name: | growth factor independent 1B transcriptional repressor |
Gene Aliases: |
BDPLT17, ZNF163B |
Location: |
Chr.9:132945545-132991697 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GFI1B | NM_004188.6 | NP_004179.3 | ||
BC043371.1 | AAH43371.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831743 | Chr.9:132820292 - 132982548 on Build GRCh38 | Loss | MIR548AW GFI1B AK8 TSC1 SPACA9 |
esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion | MIR3689C VAV2 FCN1 SNORD141A LOC100996574 MIR4669 LOC401557 PPP1R26-AS1 OLFM1 MIR548AW LOC105376306 SLC2A6 MED22 GBGT1 SURF6 DBH-AS1 SNORD24 SARDH RALGDS MIR6877 COL5A1-AS1 ADAMTSL2 REXO4 RXRA STKLD1 C9orf116 MIR3689F BRD3 RNU6ATAC SNORD36B SURF1 C9orf62 CELP MIR3689E TMEM8C LOC101448202 ADAMTS13 GTF3C5 SURF2 FCN2 SNORD36C CACFD1 WDR5 AK8 PPP1R26 LINC00094 LOC101928193 ABO SNORD36A COL5A1 CEL MIR3689D2 FAM163B RPL7A MIR3689B LOC100130548 SURF4 LCN1 GFI1B MIR3689A TSC1 MIR3689D1 OBP2B LOC101928525 MRPS2 SPACA9 DBH |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |