Assay Details
Target Gene Details
Entrez Gene ID: | 687 |
Gene Name: | Kruppel like factor 9 |
Gene Aliases: |
BTEB, BTEB1 |
Location: |
Chr.9:70384597-70414657 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLF9 | NM_001206.2 | NP_001197.1 | ||
AB451404.1 | ||||
AK313631.1 | ||||
BC069431.1 | AAH69431.1 | |||
BC074879.2 | AAH74879.1 | |||
BC074880.2 | AAH74880.1 | |||
D31716.1 | BAA06524.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428219 | Chr.9:70391353 - 70577619 on Build GRCh38 | Loss | TRPM3 KLF9 |
nsv1047862 | Chr.9:70131959 - 70523371 on Build GRCh38 | Gain | SMC5 MAMDC2 KLF9 SMC5-AS1 MAMDC2-AS1 |
nsv831615 | Chr.9:70237789 - 70404776 on Build GRCh38 | Gain | SMC5 KLF9 SMC5-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |