Assay Details
Target Gene Details
Entrez Gene ID: | 26468 |
Gene Name: | LIM homeobox 6 |
Gene Aliases: |
LHX6.1 |
Location: |
Chr.9:122185626-122228893 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LHX6 | XM_011518521.2 | XP_011516823.1 | ||
XM_011518522.2 | XP_011516824.1 |
Target Gene Details
Entrez Gene ID: | 254956 |
Gene Name: | MORN repeat containing 5 |
Gene Aliases: |
C9orf113, C9orf18 |
Location: |
Chr.9:122159661-122200088 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MORN5 | NM_001286828.1 | NP_001273757.1 | ||
NM_198469.3 | NP_940871.2 | |||
XM_005251878.3 | XP_005251935.1 | |||
XM_011518458.1 | XP_011516760.1 | |||
AI026684.1 | ||||
AI560944.1 | ||||
AK128877.1 | BAC87653.1 | |||
BC133690.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831715 | Chr.9:122066612 - 122226720 on Build GRCh38 | Gain | MIR4478 MORN5 NDUFA8 LHX6 TTLL11 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |