Assay Details
Target Gene Details
Entrez Gene ID: | 2776 |
Gene Name: | G protein subunit alpha q |
Gene Aliases: |
CMC1, G-ALPHA-q, GAQ, SWS |
Location: |
Chr.9:77716274-78031449 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNAQ | NM_002072.4 | NP_002063.2 | ||
XM_017014628.1 | XP_016870117.1 | |||
AF011496.1 | AAB64301.1 | |||
AF329284.1 | AAG61117.1 | |||
AF493896.1 | AAM12610.1 | |||
AK298579.1 | ||||
BC057777.1 | AAH57777.1 | |||
BC067850.1 | ||||
BC069520.1 | AAH69520.1 | |||
BC075096.2 | AAH75096.1 | |||
BC075097.2 | AAH75097.1 | |||
JN184778.1 | ||||
L76256.1 | AAB39498.1 | |||
U40038.1 | AAC50363.1 | |||
U43083.1 | AAB06875.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)