Assay Details
Target Gene Details
Entrez Gene ID: | 100526694 |
Gene Name: | MSANTD3-TMEFF1 readthrough |
Gene Aliases: |
C9orf3-TMEFF1, C9orf30-TMEFF1 |
Location: |
Chr.9:100441906-100577636 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MSANTD3-TMEFF1 | NM_001198812.1 | NP_001185741.1 | ||
AL831919.1 | CAD38580.2 |
Target Gene Details
Entrez Gene ID: | 8577 |
Gene Name: | transmembrane protein with EGF like and two follistatin like domains 1 |
Gene Aliases: |
C9orf2, CT120.1, H7365, TR-1 |
Location: |
Chr.9:100473238-100577636 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEFF1 | NM_003692.4 | NP_003683.2 | ||
BC035056.1 | AAH35056.1 | |||
DN602026.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv825012 | Chr.9:100389923 - 100493999 on Build GRCh38 | Gain | MSANTD3-TMEFF1 MSANTD3 TMEFF1 LOC105376177 |
nsv614955 | Chr.9:100419314 - 100498945 on Build GRCh38 | Loss | MSANTD3-TMEFF1 MSANTD3 TMEFF1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |