Assay Details
Target Gene Details
Entrez Gene ID: | 10044 |
Gene Name: | SH2 domain containing 3C |
Gene Aliases: |
CHAT, NSP3, PRO34088, SHEP1 |
Location: |
Chr.9:127738317-127778769 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SH2D3C | NM_001142531.1 | NP_001136003.1 | ||
NM_001142532.1 | NP_001136004.1 | |||
NM_001142533.1 | NP_001136005.1 | |||
NM_001142534.1 | NP_001136006.1 | |||
NM_001252334.1 | NP_001239263.1 | |||
NM_005489.3 | NP_005480.2 | |||
NM_170600.2 | NP_733745.1 | |||
XM_005251639.1 | XP_005251696.1 | |||
XM_011518114.1 | XP_011516416.1 | |||
XM_011518115.1 | XP_011516417.1 | |||
XM_011518117.2 | XP_011516419.1 | |||
XM_017014174.1 | XP_016869663.1 | |||
AF124251.1 | AAD28246.1 | |||
AK056068.1 | ||||
AK096983.1 | ||||
AK128287.1 | ||||
AK291393.1 | ||||
AK292041.1 | ||||
AK292718.1 | ||||
AK294386.1 | ||||
AY358089.1 | AAQ88456.1 | |||
AY358440.1 | AAQ89948.1 | |||
BC027962.1 | AAH27962.1 | |||
BC032365.1 | AAH32365.1 | |||
BX647905.1 | CAI46101.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv12879n54 | Chr.9:127731234 - 127824409 on Build GRCh38 | Loss | LOC102723566 MIR2861 CDK9 TTC16 ENG SH2D3C FPGS MIR3960 TOR2A |
nsv825094 | Chr.9:127742027 - 127802620 on Build GRCh38 | Gain | MIR2861 CDK9 SH2D3C MIR3960 |
nsv615371 | Chr.9:127735841 - 127787855 on Build GRCh38 | Loss | MIR2861 CDK9 SH2D3C MIR3960 |
nsv471323 | Chr.9:127727464 - 127845239 on Build GRCh38 | Loss | LOC102723566 MIR2861 CDK9 TTC16 ENG SH2D3C FPGS MIR3960 TOR2A |
nsv466571 | Chr.9:127707924 - 127774438 on Build GRCh38 | Loss | TTC16 PTRH1 SH2D3C CFAP157 TOR2A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |