Assay Details
Target Gene Details
Entrez Gene ID: | 48 |
Gene Name: | aconitase 1 |
Gene Aliases: |
ACONS, HEL60, IREB1, IREBP, IREBP1, IRP1 |
Location: |
Chr.9:32384603-32450836 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACO1 | NM_001278352.1 | NP_001265281.1 | ||
NM_002197.2 | NP_002188.1 | |||
XM_005251476.1 | XP_005251533.1 | |||
AB209480.1 | BAD92717.1 | |||
AF261088.1 | AAF99681.1 | |||
AK057904.1 | ||||
AK308752.1 | ||||
BP287929.1 | ||||
DA103947.1 | ||||
DA409113.1 | ||||
DA780353.1 | ||||
Z11559.1 | CAA77651.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1020675 | Chr.9:32323273 - 32391249 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)