Assay Details
Target Gene Details
Entrez Gene ID: | 91283 |
Gene Name: | Myb/SANT DNA binding domain containing 3 |
Gene Aliases: |
C9orf30, L8 |
Location: |
Chr.9:100427213-100451734 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MSANTD3 | NM_001198805.1 | NP_001185734.1 | ||
NM_001198806.1 | NP_001185735.1 | |||
NM_001198807.1 | NP_001185736.1 | |||
NM_080655.2 | NP_542386.1 | |||
AK314924.1 | ||||
BC008993.1 | AAH08993.1 | |||
BI596498.1 | ||||
BI600290.1 | ||||
BM914752.1 | ||||
CN366267.1 | ||||
DA424604.1 | ||||
DR159535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv825012 | Chr.9:100389923 - 100493999 on Build GRCh38 | Gain | LOC105376177 TMEFF1 MSANTD3-TMEFF1 MSANTD3 |
nsv614955 | Chr.9:100419314 - 100498945 on Build GRCh38 | Loss | TMEFF1 MSANTD3-TMEFF1 MSANTD3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |