Assay Details
Target Gene Details
Entrez Gene ID: | 100507299 |
Gene Name: | SMC5 antisense RNA 1 (head to head) |
Gene Aliases: |
MAMDC2-AS2 |
Location: |
Chr.9:70216059-70258874 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMC5-AS1 | NR_039990.1 | |||
AK124136.1 | ||||
CR936772.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv6555 | Chr.9:70225054 - 70259811 on Build GRCh38 | Insertion | SMC5-AS1 SMC5 MAMDC2 |
nsv1047862 | Chr.9:70131959 - 70523371 on Build GRCh38 | Gain | KLF9 MAMDC2-AS1 SMC5-AS1 SMC5 MAMDC2 |