Assay Details
Target Gene Details
Entrez Gene ID: | 6418 |
Gene Name: | SET nuclear proto-oncogene |
Gene Aliases: |
2PP2A, I2PP2A, IGAAD, IPP2A2, PHAPII, TAF-I, TAF-IBETA |
Location: |
Chr.9:128683432-128696396 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SET | NM_001122821.1 | NP_001116293.1 | ||
NM_001248000.1 | NP_001234929.1 | |||
XM_017015013.1 | XP_016870502.1 | |||
XM_017015014.1 | XP_016870503.1 | |||
XM_017015015.1 | XP_016870504.1 | |||
AK300794.1 | ||||
BU543146.1 | ||||
CB129868.1 | ||||
D45198.1 | BAA08139.1 | |||
JF729356.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv12882n54 | Chr.9:128663931 - 128690219 on Build GRCh38 | Loss | SET |
nsv615393 | Chr.9:128681588 - 128690033 on Build GRCh38 | Gain | SET |
nsv825097 | Chr.9:128648939 - 128702348 on Build GRCh38 | Loss | SET WDR34 |
nsv825106 | Chr.9:128686364 - 128692543 on Build GRCh38 | Gain | SET |
nsv482132 | Chr.9:128683655 - 128696396 on Build GRCh38 | Gain | SET |
dgv1237n67 | Chr.9:128662299 - 128700553 on Build GRCh38 | Gain | SET |
nsv615390 | Chr.9:128654590 - 128691724 on Build GRCh38 | Gain | SET WDR34 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |